2-(6-Chlor-3-pyridazinyl)-2-(4-methylphenyl)acetonitril
Lieferant: BIONET RESEARCH
Synonyme:
6-Chloro-alpha-(4-methylphenyl)-3-pyridazineacetonitrile
7E-003.500
7E-003.1
7E-003.5
7E-003.10
BIOT7E-003.500EA
0
EUR
BIOT7E-003.500
BIOT7E-003.1
BIOT7E-003.5
BIOT7E-003.10
2-(6-Chlor-3-pyridazinyl)-2-(4-methylphenyl)acetonitril
2-(6-Chlor-3-pyridazinyl)-2-(4-methylphenyl)acetonitril
Formel:
C₁₃H₁₀ClN₃ Molecular Weight: 243,70 g/mol |
MDL:
MFCD00214777 CAS-Nummer: 339008-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...