2-(6-Chlor-3-pyridazinyl)-2-phenylacetonitril

Lieferant: BIONET RESEARCH

Synonyme: 6-Chloro-α-phenyl-3-pyridazineacetonitrile, 2-(6-Chloropyridazin-3-yl)-2-phenylacetonitrile

5D-128.500 5D-128.1 5D-128.5 5D-128.10
BIOT5D-128.500EA 0 EUR
BIOT5D-128.500 BIOT5D-128.1 BIOT5D-128.5 BIOT5D-128.10
2-(6-Chlor-3-pyridazinyl)-2-phenylacetonitril
2-(6-Chlor-3-pyridazinyl)-2-phenylacetonitril
Formel: C₁₂H₈ClN₃
Molecular Weight: 229.6650 g/mol
MDL: MFCD00140699
CAS-Nummer: 73535-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR