2-(6-Chlor-3-pyridazinyl)-2-(4-methoxyphenyl)acetonitril

Lieferant: BIONET RESEARCH

5D-109.500 5D-109.1 5D-109.5 5D-109.10
BIOT5D-109.500EA 0 EUR
BIOT5D-109.500 BIOT5D-109.1 BIOT5D-109.5 BIOT5D-109.10
2-(6-Chlor-3-pyridazinyl)-2-(4-methoxyphenyl)acetonitril
2-(6-Chlor-3-pyridazinyl)-2-(4-methoxyphenyl)acetonitril
Formel: C₁₃H₁₀ClN₃O
MDL: MFCD00140696
CAS-Nummer: 338752-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR