2-(6-Chlor-3-pyridazinyl)-2-(2-fluorphenyl)acetonitril

Lieferant: BIONET RESEARCH

1F-011.500 1F-011.1 1F-011.5 1F-011.10
BIOT1F-011.500EA 0 EUR
BIOT1F-011.500 BIOT1F-011.1 BIOT1F-011.5 BIOT1F-011.10
2-(6-Chlor-3-pyridazinyl)-2-(2-fluorphenyl)acetonitril
2-(6-Chlor-3-pyridazinyl)-2-(2-fluorphenyl)acetonitril
Formel: C₁₂H₇ClFN₃
MDL: MFCD00202113
CAS-Nummer: 154419-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR