2-(6-Chlor-3-pyridazinyl)-2-(2-pyridyl)acetonitril

Lieferant: BIONET RESEARCH

6D-031.500 6D-031.1 6D-031.5 6D-031.10
BIOT6D-031.500EA 0 EUR
BIOT6D-031.500 BIOT6D-031.1 BIOT6D-031.5 BIOT6D-031.10
2-(6-Chlor-3-pyridazinyl)-2-(2-pyridyl)acetonitril
2-(6-Chlor-3-pyridazinyl)-2-(2-pyridyl)acetonitril
Formel: C₁₁H₇ClN₄
MDL: MFCD00140706
CAS-Nummer: 338779-25-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR