1-(6-Chlor-3-pyridazinyl)piperidin-4-on
Lieferant: COMBI-BLOCKS
Synonyme:
1-(6-Chloropyridazin-3-yl)piperidin-4-one
HC-2023-5G
HC-2023-1G
COBBHC-2023-5GEA
227
EUR
COBBHC-2023-5G
COBBHC-2023-1G
1-(6-Chlor-3-pyridazinyl)piperidin-4-on
1-(6-Chlor-3-pyridazinyl)piperidin-4-on
Formel:
C₉H₁₀ClN₃O Molecular Weight: 211,648 g/mol |
MDL:
MFCD00665150 CAS-Nummer: 303149-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...