1-(6-Chlor-3-pyridazinyl)piperidin-4-ol

Lieferant: COMBI-BLOCKS

HC-2024-5G HC-2024-25G HC-2024-1G
COBBHC-2024-5GEA 0 EUR
COBBHC-2024-5G COBBHC-2024-25G COBBHC-2024-1G
1-(6-Chlor-3-pyridazinyl)piperidin-4-ol
1-(6-Chlor-3-pyridazinyl)piperidin-4-ol
Formel: C₉H₁₂ClN₃O
MDL: MFCD05864736
CAS-Nummer: 89937-26-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR