4-(6-Chlorpyridazin-3-yl)morpholin

Lieferant: COMBI-BLOCKS

HC-6263-5G HC-6263-1G
COBBHC-6263-5GEA 359 EUR
COBBHC-6263-5G COBBHC-6263-1G
4-(6-Chlorpyridazin-3-yl)morpholin
4-(6-Chlorpyridazin-3-yl)morpholin
Formel: C₈H₁₀ClN₃O
Molecular Weight: 199,6375 g/mol
MDL: MFCD00574575
CAS-Nummer: 17259-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR