3-Chlor-6-(3,5-dimethyl-1H-pyrazol-1-yl)pyridazin

Lieferant: COMBI-BLOCKS

HC-6102-25G HC-6102-5G HC-6102-1G
COBBHC-6102-25GEA 486 EUR
COBBHC-6102-25G COBBHC-6102-5G COBBHC-6102-1G
3-Chlor-6-(3,5-dimethyl-1H-pyrazol-1-yl)pyridazin
3-Chloro-6-(3,5-dimethyl-1H-pyrazol-1-yl)pyridazine
Formel: C₉H₉ClN₄
MDL: MFCD00112281
CAS-Nummer: 29334-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR