2-(Trimethylsilylethinyl)pyridin ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2-Pyridylethynyl)trimethylsilane, 2-[(Trimethylsilyl)ethynyl]pyridine, 2-((Trimethylsilyl)ethynyl)pyridine

L11251.03 L11251.06
L11251.03EA 39.9 EUR
L11251.03 L11251.06
2-(Trimethylsilylethinyl)pyridin ≥97%
2-(Trimethylsilylethinyl)pyridin

Purity: 97% CAS No.: 86521-05-3 Molecular Formula: C10H13NSi Formula Weight: 175.31 Storage Temperature: 2°C to 8°C Refractive Index: 1.5330 Melting Point: 40-43° Boiling Point: 85-86°/1mm Beilstein Reference: 4176916 MDL No.: MFCD00066349

Formel: C₁₀H₁₃NSi
Molecular Weight: 175,31 g/mol
Siedepunkt: 85…86 °C (1 mmHg)
Schmelzpunkt: 40…43 °C
MDL: MFCD00066349
CAS-Nummer: 86521-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR