2-(Trimethylsilylethinyl)pyridin

Lieferant: CHEMPUR SILANE

Synonyme: (2-Pyridylethynyl)trimethylsilane, 2-[(Trimethylsilyl)ethynyl]pyridine, 2-((Trimethylsilyl)ethynyl)pyridine

SILS19425.5 SILS19425.25
SILNSILS19425.5EA 0 EUR
SILNSILS19425.5 SILNSILS19425.25
2-(Trimethylsilylethinyl)pyridin
2-(Trimethylsilylethinyl)pyridin
Formel: C₁₀H₁₃NSi
Molecular Weight: 175,31 g/mol
MDL: MFCD00066349
CAS-Nummer: 86521-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR