3-Brom-2-methyl-6-(4-morpholino)pyridin
Lieferant: COMBI-BLOCKS
PY-1217-5G
PY-1217-25G
PY-1217-1G
COBBPY-1217-5GEA
884
EUR
COBBPY-1217-5G
COBBPY-1217-25G
COBBPY-1217-1G
3-Brom-2-methyl-6-(4-morpholino)pyridin
3-Brom-2-methyl-6-(4-morpholino)pyridin
Formel:
C₁₀H₁₃BrN₂O Molecular Weight: 257,127 g/mol |
MDL:
MFCD13195775 CAS-Nummer: 1199773-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...