2-(4-Boc-piperazino)-2-(2-fluorphenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck

12234-250MG-F 12234-1G-F
SIAL12234-250MG-FEA 173 EUR
SIAL12234-250MG-F SIAL12234-1G-F
2-(4-Boc-piperazino)-2-(2-fluorphenyl)essigsäure, Sigma-Aldrich®
2-(4-Boc-piperazino)-2-(2-fluorphenyl)essigsäure
Formel: C₁₇H₂₃FN₂O₄
MDL: MFCD06659333
CAS-Nummer: 444892-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR