Trimethylsilylisothiocyanat

Lieferant: CHEMPUR SILANE

SILS19600.5 SILS19600.25
SILNSILS19600.5EA 0 EUR
SILNSILS19600.5 SILNSILS19600.25
Trimethylsilylisothiocyanat
Trimethylsilylisothiocyanat
Formel: C₄H₉NSSi
Molecular Weight: 131.27 g/mol
MDL: MFCD00004797
CAS-Nummer: 2290-65-5
EINECS: 218-929-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR