2-(Cyclopropylmethoxy)-5-pyridinboronsäurepinakolester ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(Cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester
H50139.03
H50139.MD
H50139.03EA
121
EUR
H50139.03
H50139.MD
2-(Cyclopropylmethoxy)-5-pyridinboronsäurepinakolester ≥97%
2-(Cyclopropylmethoxy)-5-pyridinboronsäurepinakolester
Purity: 97%
CAS No.: 947191-69-7
Molecular Formula: C15H22BNO3
Formula Weight: 275.16
MDL No.: MFCD07781184
Formel:
C₁₅H₂₂BNO₃ Molecular Weight: 275.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781184 CAS-Nummer: 947191-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...