2-(Cyclopropylmethoxy)-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-(Cyclopropylmethoxy)pyridin-3-yl)boronic acid

BB-8714-1G BB-8714-250M
COBBBB-8714-1GEA 204 EUR
COBBBB-8714-1G COBBBB-8714-250M
2-(Cyclopropylmethoxy)-5-pyridinboronsäure
2-(Cyclopropylmethoxy)-5-pyridinboronsäure
Formel: C₉H₁₂BNO₃
Molecular Weight: 193.0100 g/mol
MDL: MFCD12964560
CAS-Nummer: 1028749-31-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR