5-Brom-1-ethylbenzimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
5-Bromo-1-ethyl-1H-benzo[d]imidazole
HI-1571-1G
HI-1571-5G
HI-1571-250M
COBBHI-1571-1GEA
497
EUR
COBBHI-1571-1G
COBBHI-1571-5G
COBBHI-1571-250M
5-Brom-1-ethylbenzimidazol
5-Bromo-1-ethylbenzimidazole
Formel:
C₉H₉BrN₂ Molecular Weight: 225,852 g/mol |
MDL:
MFCD11868705 CAS-Nummer: 23073-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...