4-Brom-6-chlor-2-methylbenzimidazol
Lieferant: COMBI-BLOCKS
HC-3253-1G
HC-3253-5G
HC-3253-250M
COBBHC-3253-1GEA
208
EUR
COBBHC-3253-1G
COBBHC-3253-5G
COBBHC-3253-250M
4-Brom-6-chlor-2-methylbenzimidazol
4-Bromo-6-chloro-2-methylbenzimidazole
Formel:
C₈H₆BrClN₂ |
MDL:
MFCD15143561 CAS-Nummer: 1215205-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...