5-Cyan-1-methylbenzimidazol

Lieferant: COMBI-BLOCKS

IN-0211-5G IN-0211-1G
COBBIN-0211-5GEA 884 EUR
COBBIN-0211-5G COBBIN-0211-1G
5-Cyan-1-methylbenzimidazol
5-Cyano-1-methylbenzimidazole
Formel: C₉H₇N₃
MDL: MFCD09027482
CAS-Nummer: 53484-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR