6,7-Dimethoxy-1,2,3,4-tetrahydroisochinolin Hydrochlorid ≥97%
Lieferant: Thermo Scientific
Achtung
B22469.06 B22469.14 B22469.22
B22469.06EA 37.2 EUR
B22469.06 B22469.14 B22469.22
6,7-Dimethoxy-1,2,3,4-tetrahydroisochinolin Hydrochlorid ≥97%
6,7-Dimethoxy-1,2,3,4-tetrahydroisochinolin Hydrochlorid
Purity: 97% CAS No.: 2328-12-3 Molecular Formula: C11H15NO2.HCl Formula Weight: 229.71 Melting Point: 256-260° Beilstein Reference: 3634126
Formel: C₁₁H₁₆ClNO₂
Molecular Weight: 229,71 g/mol
Schmelzpunkt: 256…260 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012744
CAS-Nummer: 2328-12-3