5-Brom-7-chlorchinoxalin
Lieferant: COMBI-BLOCKS
HC-3252-5G
HC-3252-1G
HC-3252-250M
COBBHC-3252-5GEA
0
EUR
COBBHC-3252-5G
COBBHC-3252-1G
COBBHC-3252-250M
5-Brom-7-chlorchinoxalin
5-Bromo-7-chloroquinoxaline
Formel:
C₈H₄BrClN₂ |
MDL:
MFCD15143560 CAS-Nummer: 1215205-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...