6,7-Dichlorchinoxalin-2,3-dion

Lieferant: COMBI-BLOCKS

Synonyme: 6,7-Dichloroquinoxaline-2,3(1H,4H)-dione, 6,7-Dichloro-1,4-dihydroquinoxaline-2,3-dione

HC-6206-25G HC-6206-5G HC-6206-1G
COBBHC-6206-25GEA 889 EUR
COBBHC-6206-25G COBBHC-6206-5G COBBHC-6206-1G
6,7-Dichlorchinoxalin-2,3-dion
6,7-Dichlorchinoxalin-2,3-dion
Formel: C₈H₄Cl₂N₂O₂
Molecular Weight: 231,0356 g/mol
MDL: MFCD00078575
CAS-Nummer: 25983-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR