6-Cyan-7-nitrochinoxalin-2,3-dion ≥98% (durch HPLC), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
7-Nitro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile, 6-Cyano-7-nitroquinoxaline-2,3(1H,4H)-dione, CNQX
M-Clarity™ Qualitätsstufe = MQ100
C127-25MG
C127-50MG
C127-5MG
SIALC127-25MGEA
589
EUR
SIALC127-25MG
SIALC127-50MG
SIALC127-5MG
6-Cyan-7-nitrochinoxalin-2,3-dion ≥98% (durch HPLC), Sigma-Aldrich®
6-Cyan-7-nitrochinoxalin-2,3-dion
Formel:
C₉H₄N₄O₄ Molecular Weight: 232,16 g/mol |
MDL:
MFCD00069232 CAS-Nummer: 115066-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...