Methylchinoxalin-6-carboxylat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 6-Quinoxalinecarboxylate

11X-0803.10 11X-0803.5 11X-0803.1 11X-0803.500
BIOT11X-0803.10EA 0 EUR
BIOT11X-0803.10 BIOT11X-0803.5 BIOT11X-0803.1 BIOT11X-0803.500
Methylchinoxalin-6-carboxylat
Methylchinoxalin-6-carboxylat
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188.19 g/mol
MDL: MFCD00102618
CAS-Nummer: 23088-23-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR