Methylchinoxalin-6-carboxylat
Lieferant: Thermo Scientific
Synonyme:
Methyl 6-Quinoxalinecarboxylate
RDP00059DA
RDP00059EA
MAYBRDP00059DAEA
0
EUR
MAYBRDP00059DA
MAYBRDP00059EA
Methylchinoxalin-6-carboxylat
Methylchinoxalin-6-carboxylat
Formel:
C₁₀H₈N₂O₂ Molecular Weight: 188,19 g/mol |
MDL:
MFCD00102618 CAS-Nummer: 23088-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...