6-Chinoxalincarbonsäure 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Quinoxaline-6-carboxylic acid

H33157.03 H33157.06 H33157.14
H33157.03EA 58.8 EUR
H33157.03 H33157.06 H33157.14
6-Chinoxalincarbonsäure 95%
6-Quinoxalinecarboxylic acid

Purity: 95% CAS No.: 6925-00-4 Molecular Formula: C9H6N2O2 Formula Weight: 174.16 Melting Point: 248-251° MDL No.: MFCD01365834

Formel: C₉H₆N₂O₂
Molecular Weight: 174.16 g/mol
Schmelzpunkt: 248…251 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01365834
CAS-Nummer: 6925-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR