6-Chinoxalincarbonsäure

Lieferant: BIONET RESEARCH

Synonyme: Quinoxaline-6-carboxylic acid

11X-0826.5 11X-0826.10 11X-0826.500 11X-0826.1
BIOT11X-0826.5EA 0 EUR
BIOT11X-0826.5 BIOT11X-0826.10 BIOT11X-0826.500 BIOT11X-0826.1
6-Chinoxalincarbonsäure
6-Quinoxalinecarboxylic acid
Formel: C₉H₆N₂O₂
Molecular Weight: 174.16 g/mol
MDL: MFCD01365834
CAS-Nummer: 6925-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR