6-Chinoxalincarbonsäure
Lieferant: Thermo Scientific
Synonyme:
Quinoxaline-6-carboxylic acid
CC11701DA
CC11701EA
MAYBCC11701DAEA
0
EUR
MAYBCC11701DA
MAYBCC11701EA
6-Chinoxalincarbonsäure
6-Quinoxalinecarboxylic acid
Formel:
C₉H₆N₂O₂ Molecular Weight: 174,16 g/mol |
MDL:
MFCD01365834 CAS-Nummer: 6925-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...