5,7-Difluorchinoxalin

Lieferant: COMBI-BLOCKS

HC-3265-5G HC-3265-1G
COBBHC-3265-5GEA 303 EUR
COBBHC-3265-5G COBBHC-3265-1G
5,7-Difluorchinoxalin
5,7-Difluoroquinoxaline
Formel: C₈H₄F₂N₂
MDL: MFCD15143566
CAS-Nummer: 1215205-81-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR