Chinoxalin-6-amin

Lieferant: BIONET RESEARCH

Synonyme: 6-Aminoquinoxaline

5R-0605.500 5R-0605.1 5R-0605.5 5R-0605.10
BIOT5R-0605.500EA 0 EUR
BIOT5R-0605.500 BIOT5R-0605.1 BIOT5R-0605.5 BIOT5R-0605.10
Chinoxalin-6-amin
Chinoxalin-6-amin
Formel: C₈H₇N₃
Molecular Weight: 145.1600 g/mol
MDL: MFCD00462821
CAS-Nummer: 6298-37-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR