6-Chlor-7-methylchinoxalin
Lieferant: COMBI-BLOCKS
HC-3257-5G
HC-3257-1G
COBBHC-3257-5GEA
451
EUR
COBBHC-3257-5G
COBBHC-3257-1G
6-Chlor-7-methylchinoxalin
6-Chloro-7-methylquinoxaline
Formel:
C₉H₇ClN₂ |
MDL:
MFCD15143565 CAS-Nummer: 646504-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...