2-Chlor-3-(trifluormethyl)chinoxalin

Lieferant: CHEMPUR

BTB13029.250 BTB13029.1
CHMPBTB13029.250EA 0 EUR
CHMPBTB13029.250 MAYBBTB13029.1
2-Chlor-3-(trifluormethyl)chinoxalin
2-Chloro-3-(trifluoromethyl)quinoxaline
Formel: C₉H₄ClF₃N₂
Molecular Weight: 232,59 g/mol
MDL: MFCD00243891
CAS-Nummer: 254732-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR