2,3-Dimethylchinoxalin-6-amin
Lieferant: BIONET RESEARCH
5R-0644.500
5R-0644.1
5R-0644.5
5R-0644.10
BIOT5R-0644.500EA
0
EUR
BIOT5R-0644.500
BIOT5R-0644.1
BIOT5R-0644.5
BIOT5R-0644.10
2,3-Dimethylchinoxalin-6-amin
2,3-Dimethylchinoxalin-6-amin
Formel:
C₁₀H₁₁N₃ |
MDL:
MFCD01759944 CAS-Nummer: 7576-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...