2,3-Dimethyl-6-nitrochinoxalin

Lieferant: BIONET RESEARCH

1R-0607.500 1R-0607.1 1R-0607.5 1R-0607.10
BIOT1R-0607.500EA 0 EUR
BIOT1R-0607.500 BIOT1R-0607.1 BIOT1R-0607.5 BIOT1R-0607.10
2,3-Dimethyl-6-nitrochinoxalin
2,3-Dimethyl-6-nitroquinoxaline
Formel: C₁₀H₉N₃O₂
MDL: MFCD00205436
CAS-Nummer: 2942-03-2
EINECS: 219-162-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR