4-(Methoxycarbonyl)-2-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-4-carboxylate
PN-5523-1G
PN-5523-250M
COBBPN-5523-1GEA
1110
EUR
COBBPN-5523-1G
COBBPN-5523-250M
4-(Methoxycarbonyl)-2-indolboronsäurepinakolester
4-(Methoxycarbonyl)-2-indolboronsäurepinakolester
Formel:
C₁₆H₂₀BNO₄ Molecular Weight: 301,1500 g/mol |
MDL:
MFCD11858381 CAS-Nummer: 1072811-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...