5-(Methoxycarbonyl)-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-5-carboxylate

FM-2506-1G FM-2506-250M
COBBFM-2506-1GEA 1110 EUR
COBBFM-2506-1G COBBFM-2506-250M
5-(Methoxycarbonyl)-2-indolboronsäurepinakolester
5-(Methoxycarbonyl)-2-indolboronsäurepinakolester
Formel: C₁₆H₂₀BNO₄
Molecular Weight: 301,15 g/mol
MDL: MFCD11858383
CAS-Nummer: 1256358-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR