6-(Methoxycarbonyl)-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-6-carboxylate

PN-5577-1G PN-5577-250M
COBBPN-5577-1GEA 747 EUR
COBBPN-5577-1G COBBPN-5577-250M
6-(Methoxycarbonyl)-2-indolboronsäurepinakolester
6-(Methoxycarbonyl)-2-indolboronsäurepinakolester
Formel: C₁₆H₂₀BNO₄
Molecular Weight: 301,1453 g/mol
MDL: MFCD11858368
CAS-Nummer: 1256359-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR