7-(Methoxycarbonyl)-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-7-carboxylate

PN-5417-1G PN-5417-250M
COBBPN-5417-1GEA 585 EUR
COBBPN-5417-1G COBBPN-5417-250M
7-(Methoxycarbonyl)-2-indolboronsäurepinakolester
7-(Methoxycarbonyl)-2-indolboronsäurepinakolester
Formel: C₁₆H₂₀BNO₄
Molecular Weight: 301,15 g/mol
MDL: MFCD11504970
CAS-Nummer: 1150271-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR