1-Boc-2-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate
PN-2009-5G
PN-2009-25G
PN-2009-1G
COBBPN-2009-5GEA
595
EUR
COBBPN-2009-5G
COBBPN-2009-25G
COBBPN-2009-1G
1-Boc-2-indolboronsäurepinakolester
1-Boc-2-indolboronsäurepinakolester
Formel:
C₁₉H₂₆BNO₄ Molecular Weight: 343.2251 g/mol |
MDL:
MFCD09838985 CAS-Nummer: 1072944-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...