1-Boc-3-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

PN-6187-1G PN-6187-250M
COBBPN-6187-1GEA 147 EUR
COBBPN-6187-1G COBBPN-6187-250M
1-Boc-3-indolboronsäurepinakolester
1-Boc-3-indolboronsäurepinakolester
Formel: C₁₉H₂₆BNO₄
Molecular Weight: 343.2251 g/mol
MDL: MFCD12407262
CAS-Nummer: 942070-45-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR