1-Boc-5-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate
PN-8864-5G
PN-8864-1G
COBBPN-8864-5GEA
181
EUR
COBBPN-8864-5G
COBBPN-8864-1G
1-Boc-5-indolboronsäurepinakolester
1-Boc-5-indolboronsäurepinakolester
Formel:
C₁₉H₂₆BNO₄ Molecular Weight: 343.2251 g/mol |
MDL:
MFCD05663895 CAS-Nummer: 777061-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...