1-Boc-5-indolboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

640387-1G 640387-5G
SIAL640387-1GEA 56.2 EUR
SIAL640387-1G SIAL640387-5G
1-Boc-5-indolboronsäurepinakolester, Sigma-Aldrich®
1-Boc-5-indolboronsäurepinakolester
Formel: C₁₉H₂₆BNO₄
Molecular Weight: 343.2251 g/mol
MDL: MFCD05663895
CAS-Nummer: 777061-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR