1-Boc-5-methoxy-3-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl 5-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate
PN-6301-1G
PN-6301-250M
COBBPN-6301-1GEA
742
EUR
COBBPN-6301-1G
COBBPN-6301-250M
1-Boc-5-methoxy-3-indolboronsäurepinakolester
1-Boc-5-methoxy-3-indolboronsäurepinakolester
Formel:
C₂₀H₂₈BNO₅ Molecular Weight: 373.2510 g/mol |
MDL:
MFCD16295115 CAS-Nummer: 1256359-99-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...