1-Boc-6-methoxy-3-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl 6-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate
PN-6302-1G
PN-6302-250M
COBBPN-6302-1GEA
742
EUR
COBBPN-6302-1G
COBBPN-6302-250M
1-Boc-6-methoxy-3-indolboronsäurepinakolester
1-Boc-6-methoxy-3-indolboronsäurepinakolester
Formel:
C₂₀H₂₈BNO₅ Molecular Weight: 373.2510 g/mol |
MDL:
MFCD14155740 CAS-Nummer: 1256360-00-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...