1-Boc-7-methoxy-3-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl 7-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate
PN-6220-1G
PN-6220-250M
COBBPN-6220-1GEA
1110
EUR
COBBPN-6220-1G
COBBPN-6220-250M
1-Boc-7-methoxy-3-indolboronsäurepinakolester
1-Boc-7-methoxy-3-indolboronsäurepinakolester
Formel:
C₂₀H₂₈BNO₅ Molecular Weight: 373.2500 g/mol |
MDL:
MFCD14155739 CAS-Nummer: 1218790-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...