1-Boc-7-(methoxycarbonyl)-3-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-tert-Butyl 7-methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,7-dicarboxylate

PN-6314-1G PN-6314-250M
COBBPN-6314-1GEA 1110 EUR
COBBPN-6314-1G COBBPN-6314-250M
1-Boc-7-(methoxycarbonyl)-3-indolboronsäurepinakolester
1-Boc-7-(methoxycarbonyl)-3-indolboronsäurepinakolester
Formel: C₂₁H₂₈BNO₆
Molecular Weight: 401,2600 g/mol
MDL: MFCD16295116
CAS-Nummer: 1256360-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR