4-Methoxy-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Methoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

FM-2508-1G FM-2508-250M
COBBFM-2508-1GEA 612 EUR
COBBFM-2508-1G COBBFM-2508-250M
4-Methoxy-2-indolboronsäurepinakolester
4-Methoxy-2-indolboronsäurepinakolester
Formel: C₁₅H₂₀BNO₃
Molecular Weight: 273.1352 g/mol
MDL: MFCD11858387
CAS-Nummer: 1072811-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR