6-Methoxy-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 6-Methoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

PN-6266-1G PN-6266-250M
COBBPN-6266-1GEA 1110 EUR
COBBPN-6266-1G COBBPN-6266-250M
6-Methoxy-2-indolboronsäurepinakolester
6-Methoxy-2-indolboronsäurepinakolester
Formel: C₁₅H₂₀BNO₃
Molecular Weight: 273,1400 g/mol
MDL: MFCD16295111
CAS-Nummer: 1256359-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR