5-Methoxy-2,7-diindolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
5-Methoxy-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
PN-6421-1G
PN-6421-250M
COBBPN-6421-1GEA
1110
EUR
COBBPN-6421-1G
COBBPN-6421-250M
5-Methoxy-2,7-diindolboronsäurepinakolester
5-Methoxy-2,7-diindolboronsäurepinakolester
Formel:
C₂₁H₃₁B₂NO₅ Molecular Weight: 399,0966 g/mol |
MDL:
MFCD15474883 CAS-Nummer: 1256360-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...