4-Brom-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

FM-2507-1G FM-2507-250M
COBBFM-2507-1GEA 595 EUR
COBBFM-2507-1G COBBFM-2507-250M
4-Brom-2-indolboronsäurepinakolester
4-Brom-2-indolboronsäurepinakolester
Formel: C₁₄H₁₇BBrNO₂
MDL: MFCD11858385
CAS-Nummer: 1256358-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR